Thermo Scientific™
Dionex™ PC10 Postcolumn Delivery System Reagent Organizer
 Related applications:
Industrial Chromatography
Use the Thermo Scientific™ Dionex™ PC10 Postcolumn Delivery System Reagent Organizer as a replacement part or spare part for the Dionex™ PC10 Postcolumn Pneumatic Delivery Package. The Dionex™ PC10 provides pulse-free reagent flow in applications with high background absorbances, such as trace-level transition metal analysis using post-column reaction with 4-(2-pyridylazo)resorcinol (PAR).
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
The PC10 Postcolumn Pneumatic Delivery System ensures a constant, pulseless flow of reagent. It assures optimal flow by applying constant pressure, eliminating the need for a pump and its associated moving parts, labor, and expense.
Documents
Manuals & protocols