Guanidinium hydrochloride, 99.5-101.0%, white powder, GenAR™ for biotechnology, GMP, Macron Fine Chemicals™
Avantor™

Guanidinium hydrochloride, 99.5-101.0%, white powder, GenAR™ for biotechnology, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets NF specifications
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogodimQuantityPackaging
0507-F7
M0507-04
M0507-06
0507-25
0507-88
0507-F9
Número de catálogo 0507-F7
Precio (MXN)
-
This product is GMP Manufactured and meets NF specifications
This product is GMP Manufactured and meets NF specifications
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
FormularioPowder
Punto de fusión185°C
Peso molecular95.53 g/mol
Tipo de empaquetadoHDPE Flowmor package
Pureza99.5 to 101%
Cantidad12 kg
GradoBiotech
Unit SizeEach