Thermo Scientific Chemicals

Potassium heptafluoroniobate(V), 99.5%, Thermo Scientific Chemicals

Catalog number: 051110.18
50 g, Each
Thermo Scientific Chemicals

Potassium heptafluoroniobate(V), 99.5%, Thermo Scientific Chemicals

Catalog number: 051110.18
50 g, Each
Quantity
Catalog number: 051110.18
also known as 051110-18
Price (USD)
Price: 144.00
Online price: 124.65
Your price:
Quantity
-

Chemical Identifiers

CAS
16924-03-1
IUPAC Name
niobium(5+) dipotassium heptafluoride
Molecular Formula
F7K2Nb
InChI Key
FTPUNAWAGWERLA-UHFFFAOYSA-G
SMILES
[F-].[F-].[F-].[F-].[F-].[F-].[F-].[K+].[K+].[Nb+5]
Assay (unspecified)
99.5% min

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text