This product is now available for online ordering in the U.S. and Canada—Learn more »
Thermo Scientific™
Dionex™ Solvent Bottle Caps and Liners
 Related applications:
Industrial Chromatography
Choose the Thermo Scientific™ Dionex™ Solvent Bottle Caps and Liners that are used to cap 1L and 2L solvent bottles. The 1L bottle is for use on the Dionex™ ASE™ Accelerated Solvent Extractor and Thermo Scientific™ AutoTrace™ SPE Solid Phase Extraction systems. The 2L bottle is for use on the Dionex ASE system. Cap liners are available in PEEK and Teflon for biocompatibility.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Dionex ASE systems reduce sample preparation from hours to minutes by performing automated solvent extraction on solid and semisolid sample matrices.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Documents
Manuals & protocols
Recommended products