Thermo Scientific Chemicals

Nickel(II) chloride hexahydrate, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 053131.18
50 g, Each
Thermo Scientific Chemicals

Nickel(II) chloride hexahydrate, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 053131.18
50 g, Each
Quantity
Catalog number: 053131.18
also known as 053131-18
Price (USD)
Price: 51.80
Online price: 44.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7791-20-0
IUPAC Name
nickel(2+) hexahydrate dichloride
Molecular Formula
Cl2H12NiO6
InChI Key
LAIZPRYFQUWUBN-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Ni++]
Appearance (Color)
Green
Form
Crystalline powder or crystals
Assay from Supplier's CofA
≥99.95% metals basis
Total Metal Impurities
≤0.05% (500 ppm)

Description

Nickel(II) chloride is used in electroplating and as a catalyst for organic conversions, for example in chemo-selective thioacetalization of aldehydes. In combination with lithium aluminum hydride, it serves as a reducing agent for alkenes, alkynes, and organic halides; it can cleave N-O bond and open epoxides. It is used to prepare a wide array of complexes since water ligands can be easily replaced by amines, thioether, ammonia, and thiolates. It is a precursor to several nickel-phosphine complexes, such as bis(triphenylphosphine)nickel(II) chloride, which are used in alkyne trimerizations, carbonylations, and as catalysts in organic reactions such as Suzuki-Miyaura cross coupling reactions as an alternative to palladium(0) catalysts. It is the precursor to acetylacetonate complex of Ni, used for producing 1,5-cyclooctadiene complex, an important reagent in organonickel chemistry. It can be used to prepare the sandwich compound nickelocene through dimethoxyethane complex of nickel chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text