Ammonium peroxydisulfate, ACS, 98.0% min
Ammonium peroxydisulfate, ACS, 98.0% min
Ammonium peroxydisulfate, ACS, 98.0% min
Thermo Scientific Chemicals

Ammonium peroxydisulfate, ACS, 98.0% min

CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 x 1 kg
Catalog number 054106.22
also known as 054106-22
Price (EUR)
32,90
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
32,90
Each
Chemical Identifiers
CAS7727-54-0
IUPAC Namediammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaH8N2O8S2
InChI KeyROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)0.04meq/g
Residue on ignition0.05%
Impurity contentChloride and chlorate (as Cl) 0.001%
Trace Metals(as Pb) 0.005%
Assay (unspecified)98.0% H₈N₂O₈S₂ min
View more
Ammonium peroxydisulfate is used as an oxidizer, a bleaching agent, in the field of photography, to make aniline dyes, electroplating and in analytical chemistry for determination of manganese.Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only