Thermo Scientific™
Dionex™ ASE™ Qualification Kit
 Related applications:
Industrial Chromatography
The Thermo Scientific™ Dionex™ ASE™ Qualification Kit is for Ion Chromatography and ASE systems.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Qualification consists of three types of system qualification: IQ - Installation Qualification, OQ - Operational Qualification, PQ - Performance Qualification;
Documents
Manuals & protocols