Thermo Scientific Chemicals

Sodium thiophosphate hydrate, Thermo Scientific Chemicals

Catalog number: 056146.14
25 g, Each
Thermo Scientific Chemicals

Sodium thiophosphate hydrate, Thermo Scientific Chemicals

Catalog number: 056146.14
25 g, Each
Quantity
Catalog number: 056146.14
also known as 056146-14
Price (USD)
Price: 357.00
Online price: 303.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10489-48-2
IUPAC Name
trisodium phosphorothioate
Molecular Formula
Na3O3PS
InChI Key
RIFYBBXGYKFBFC-UHFFFAOYSA-K
SMILES
[Na+].[Na+].[Na+].[O-]P([O-])([O-])=S
Assay (unspecified)
Na 15.5% min
Form
Crystalline aggregates

Description

Sodium thiophosphate hydrate is used in thiosulfate-assisted synthesis of a variety of diamine-CuCN complexes. In analytical chemistry, it is used as a titrant in iodometry. It finds application as an anionic surfactant in pharmaceutical preparations to assist in dispersion and an antidote to cyanide poisoning. It is utilized for the dechlorination of tap water as well as to maintain the chlorine levels in swimming pools and aquaria. It plays an important role in gold extraction. It is also used in the alkylation reaction to prepare Bunte salts viz. S-alkylthiosulfonates, which finds application in the synthesis of thioglycolic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text