사양
보정To contain
인증/적합성USP, ASTM E 438, ASTM D2709
용량(미터법)100 mL
일회용Reusable
물질Borosilicate Glass
용도Used for the determination of small quantities of solids in large volumes of liquids
스토퍼 수16
공차0 to 0.2mL in ±0.01mL, 25mL in ±1mL, >25mL in ±2mL
형상Pear
길이(미터법)160 mm
직경(미터법)58 mm
눈금0 to 0.2 mL in 0.01 mL increments, 25, 50, and 100 mL
제품 유형Centrifuge Tube
최대 RCF800 x g
Unit SizeEach