Dionex™ ASE™ Extraction Cell Frits
Actual product may vary
Dionex™ ASE™ Extraction Cell Frits
Dionex™ ASE™ Extraction Cell Frits
Thermo Scientific™
Dionex™ ASE™ Extraction Cell Frits
Use these frits to prevent plugging of sample particulates in the extraction cell of Thermo Scientific™ Dionex™ ASE™ Accelerated Solvent Extractors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialFor Use With (Equipment)Quantity
056775Stainless SteelDionex ASE 100, 150, 300 and 35050
068260DioniumDionex ASE 150 and 35010
Catalog number 056775
Price (USD)
502.00
Pack of 50
Material:
Stainless Steel
For Use With (Equipment):
Dionex ASE 100, 150, 300 and 350
Quantity:
50
Price (USD)
502.00
Pack of 50
Use these Thermo Scientific™ Dionex™ ASE™ Extraction Cell Frits to prevent plugging of sample particulates in the extraction cell. Choose from stainless steel or Dionium™ frits for your Dionex ASE system. Dionex™ ASE™ Accelerated Solvent Extractors feature extraction cells with finger-tight cell caps with compression seals for high-pressure closure and maximum ease of use.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description10 μm Frit
MaterialStainless Steel
Quantity50
For Use With (Equipment)Dionex ASE 100, 150, 300 and 350
TypeExtraction Cell Frit
Unit SizePack of 50