Dionex™ ASE™ Extraction Cell Seals
Actual product may vary
Dionex™ ASE™ Extraction Cell Seals
Dionex™ ASE™ Extraction Cell Seals
Thermo Scientific™
Dionex™ ASE™ Extraction Cell Seals
Install this seal in the end cap of the Thermo Scientific™ Dionex™ ASE™ Extraction Cell to ensure sealing between the extraction cell cap and cell body.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialFor Use With (Equipment)Quantity
056776VespelDionex ASE 150 or 350 System Extraction Cell Caps for High Temperature ExtractionsPack of 50
061687PEEKDionex ASE 100, 150, 300 and 350Pack of 50
056777VespelDionex ASE 150 or 350 System Extraction Cell Caps for High Temperature ExtractionsPack of 10
Catalog number 056776
Price (TWD)
-
Material:
Vespel
For Use With (Equipment):
Dionex ASE 150 or 350 System Extraction Cell Caps for High Temperature Extractions
Quantity:
Pack of 50
Install a new Thermo Scientific™ Dionex™ ASE™ Extraction Cell Seal to ensure sealing between the extraction cell cap and cell body. Dionex™ ASE™ Accelerated Solvent Extractors feature extraction cells with finger-tight cell caps with compression seals for high-pressure closure and maximum ease of use.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionExtraction Cell Cap Interior Seals, Vespel
MaterialVespel
QuantityPack of 50
For Use With (Equipment)Dionex ASE 150 or 350 System Extraction Cell Caps for High Temperature Extractions
TypeExtraction Cell Seal
Unit SizePack of 50