Thermo Scientific™

Dionex™ MMS Installation Kit for IC in DCR Mode

Catalog number: 056884
Thermo Scientific™

Dionex™ MMS Installation Kit for IC in DCR Mode

Catalog number: 056884
Use the Thermo Scientific™ Dionex™ MMS Installation Kit when configuring your ion chromatography (IC) system for Displacement Chemical Regeneration (DCR) mode. Conveniently sized to match a 2L or 4L eluent bottles, the kit includes a regenerant reservoir cap and all tubing and fittings to install the Thermo Scientific™ Dionex™ MMS 300 MicroMembrane Suppressor in DCR mode. This kit is compatible with Dionex™ AMMS™ Anion and CMMS™ Cation MicroMembrane Suppressors. Refer to Thermo Scientific™ Dionex™ DCR Reagents to prepare regenerants from concentrates.
 
Catalog Number
056884
Unit Size
Each
Type
MMS Installation Kit for 4 mm Columns
Price (USD)
Full specifications
For Use WithIon Chromatography Systems
OperationConfigures ion chromatography (IC) systems for Displacement Chemical Regeneration (DCR) mode.
TypeMMS Installation Kit for 4 mm Columns
IncludesOne regenerant reservoir cap, and all tubing and fittings to install the Thermo Scientific Dionex MMS 300 in. the DCR mode
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Displacement Chemical Regeneration Kit
Price (USD)
Full specifications
For Use WithIon Chromatography Systems
OperationConfigures ion chromatography (IC) systems for Displacement Chemical Regeneration (DCR) mode.
TypeDisplacement Chemical Regeneration Kit
IncludesOne regenerant reservoir cap, and all tubing and fittings to install the Thermo Scientific Dionex MMS 300 in. the DCR mode
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeTypePrice (USD)
056884Full specifications
EachMMS Installation Kit for 4 mm ColumnsRequest A Quote
For Use WithIon Chromatography Systems
OperationConfigures ion chromatography (IC) systems for Displacement Chemical Regeneration (DCR) mode.
TypeMMS Installation Kit for 4 mm Columns
IncludesOne regenerant reservoir cap, and all tubing and fittings to install the Thermo Scientific Dionex MMS 300 in. the DCR mode
Unit SizeEach
056882Full specifications
EachDisplacement Chemical Regeneration KitRequest A Quote
For Use WithIon Chromatography Systems
OperationConfigures ion chromatography (IC) systems for Displacement Chemical Regeneration (DCR) mode.
TypeDisplacement Chemical Regeneration Kit
IncludesOne regenerant reservoir cap, and all tubing and fittings to install the Thermo Scientific Dionex MMS 300 in. the DCR mode
Unit SizeEach
Showing 2 of 2
Each Dionex Displacement Chemical Regeneration Kit includes one regenerant reservoir, cap and all tubing and fittings needed to install any Dionex MMS 300 MicroMembrane Suppressor in displacement chemical regeneration mode.

Ready for Your Applications
  • Compatible with Dionex™ AMMS™ Anion and CMMS™ Cation MicroMembrane Suppressors
  • Conveniently sized to match a 2L or 4L eluent bottles
See Standards and Reagents for regenerant concentrates and solutions.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.