Product Image
Thermo Scientific™
Dionex™ OnGuard™ II P Cartridges
Remove the phenolic fraction of humic acids, tannic acids, lignins, anthocyanins, and azodyes from sample to improve IC performance. Pretreat 12 samples in parallel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
05708748/Pk.
08835812/Pk.
Catalog number 057087
Price (CLP)
-
Quantity:
48/Pk.
Remove the phenolic fraction of humic acids, tannic acids, lignins, anthocyanins, and azodyes from sample matrices. Thermo Scientific™ Dionex™ OnGuard™ II P Cartridges are an off-line sample preparation technique that uses polyvinylpyrrolidone (PVP) polymer with very high selectivity to eliminate these matrix interferences to improve IC performance and facilitate low-level ion analysis. Use Dionex OnGuard II P Cartridges singly or in series, or pretreat up to 12 samples in parallel using the optional Thermo Scientific Dionex OnGuard Sample Prep Station.
Dionex OnGuard II P Cartridges use a polyvinylpyrrolidone (PVP) polymer with very high selectivity for phenolics, azo-containing compounds, aromatic carboxylic acids, aromatic aldehydes, and iodine as the triiodide complex
Solve tough application problems
  • Boost sensitivity, lower baselines, and get better separations by removing phenolics, metals, cations, anions, hydrophobic substances, and more
  • Achieve reproducible trace-level determinations in concentrated matrices
  • Increase the lifetime of your analytical columns
  • Use for a wide range of matrices thanks to wide pH stability
  • Supports applications with choice of 1cc formats, as well as syringe barrel format for robotic processing (custom order)
Easy to Use, Leak-Free
  • Leak-free, no-channeling cartridge
  • Quick, secure Luer inlet connections
Performance Guaranteed
  • Ships with a Certificate of Analysis that verifies capacity, performance and cleanliness
Pretreat 12 Samples Simultaneously
  • Use the optional Dionex OnGuard Sample Prep Station and OnGuard Sample Pretreatment Cartridges
Specifications
DescriptionRemoves phenolics and azo dyes from samples prior to cation or anion analysis
For Use With (Application)Phenolic Fraction Removal
MaterialPolyvinylpyrrolidone Polymer
Quantity48/Pk.
Volume (Metric)1 mL
For Use With (Equipment)Dionex OnGuard Sample Prep Station
Product LineOnGuard
Unit SizeEach 1