Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Use With | IonPac AS14 Columns |
Standard Components | Sodium carbonate, Sodium bicarbonate |
Type | Eluent Concentrate |
Standard Concentration | 0.80, 0.10 X |
Concentration | 20 X |
Hazards | Irritant |
Quantity | 4 x 100 mL |
Storage | Room Temperature |
Unit Size | Each 1 |
For Use With | IonPac AS14 Columns |
Standard Components | Sodium Carbonate, Sodium Bicarbonate, Water |
Type | Eluent Concentrate |
Standard Concentration | 0.025 |
Concentration | 100 X |
Quantity | 250 mL |
Unit Size | Each 1 |
Catalog Number | Specifications | Unit Size | Quantity | Concentration | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
057557 | Each 1 | 4 x 100 mL | 20 X | Price: 175.00 Online Offer: 157.50 (ends 30-Nov-2023) Your Price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
053560 | Each 1 | 250 mL | 100 X | Price: 110.00 Online Offer: 93.50 (ends 30-Nov-2023) Your Price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|