Dionex™ Methanesulfonic Acid Cation Eluent Concentrate
Actual product may vary
Thermo Scientific™

Dionex™ Methanesulfonic Acid Cation Eluent Concentrate

For simplicity and flexibility in your cation exchange applications, ready-to-dilute Thermo Scientific™ Dionex™ Methanesulfonic Acid Cationic Eluent Concentrate is ideal.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
057562100 mL
0575584 x 100 mL
Catalog number 057562
Price (MXN)
-
Quantity:
100 mL

For simplicity and flexibility in your cation exchange applications, this ready-to-dilute Thermo Scientific™ Dionex™ Methanesulfonic Acid Cation Eluent Concentrate is ideal; just dilute to the required level with deionized water. Use this MSA cation eluent concentrate with Dionex™ IonPac™ CS12, CS12A, CS14, CS15, CS16, and CS17 columns. The 500mL concentrate is made with only high purity, reagent grade chemicals and water (18 MO-cm) and quality checked.

The methanesulfonic acid (MSA), >99% purity, is available to prepare eluents of any concentration. Methanesulfonic acid is recommended for the preparation of IonPac CS12, CS12A, CS14, CS15, CS16, CS17, and CS18 eluents. The eluent concentrates are recommended for fast, reliable eluent preparation.

  • Only high-purity, reagent-grade chemicals and water (18 MΩ-cm), quality-control checked for ionic contamination, are used
  • All standards are NIST™ traceable

Includes:

A Certificate of Analysis is provided with every standard and chelation chromatography reagent

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceNIST Traceable
Concentration20 X
DescriptionCS12 A Eluent Concentrate; 20 X; 100 mL
For Use With (Equipment)Dionex IonPac CS12, CS12A, CS14, CS15, CS16, and CS17 Columns
HazardsCorrosive
IncludesCertificate of Analysis
Purity>99%
Quantity100 mL
Standard ComponentsMethanesulfonic Acid Aqueous Solution
Standard Concentration20 X
StorageRoom Temperature
TypeEluent Concentrate
Unit SizeEach