Glycine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Glycine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™

Meets USP requirements, BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.Más información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
0582-01500 g
0582-052 kg
0582-0712 kg
0582-0950 kg
Número de catálogo 0582-01
Precio (MXN)
-
Cantidad:
500 g
Meets USP requirements, BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición181°C (1013 hPa)
CAS56-40-6
Nombre del producto químico o materialGlycine
ColorWhite
Punto de fusión83°C
Fórmula molecularNH2CH2COOH
EnvaseAmber glass bottle, wide mouth, tamper-evident cap
SinónimoAminoacetic acid H-Gly-OH
GradoMulticomponente/USP
Peso molecular (g/mol)75.07
Porcentaje de pureza98.5 to 101.5%
Cantidad500 g
pH4.0 (20°C, 0.2M aqueous solution)
Unit SizeEach