Glycine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Glycine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™

Meets USP requirements, BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
0582-0712 kg
0582-052 kg
0582-0950 kg
0582-01500 g
Catalog number 0582-07
Price (MXN)
-
Quantity:
12 kg
Meets USP requirements, BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.
Meets USP requirements, BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.
Specifications
Boiling Point181°C (1013 hPa)
CAS56-40-6
Chemical Name or MaterialGlycine
ColorWhite
Melting Point83°C
Molecular FormulaNH2CH2COOH
PackagingPolyethylene pail, screw top
SynonymAminoacetic acid
GradeMulti-Compendial/USP
Molecular Weight (g/mol)75.07
Percent Purity98.5 to 101.5%
Quantity12 kg
pH4.0 (at 20°C, 0.2M)
Unit SizeEach