Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Type | Flow Rate | Pore Size (Metric) |
---|---|---|---|
06.5101 | Prefilter Insert | 1000 L/hr. (at D p = 0.2 bar) | 1 μm |
50155506 | Prefilter Insert | 1550 L/hr. (at D p = 0.2 bar) | 5 μm |
06.5106 | Prefilter Insert | 1700 L/hr. (at D p = 0.2 bar) | 1 μm |
50155507 | Prefilter Insert | 1300 L/hr. (at D p = 0.2 bar) | 3 μm |
50155508 | Prefilter Insert | 2400 L/hr. (at D p = 0.2 bar) | 3 μm |
50155509 | Prefilter Insert | 3200 L/hr. (at D p = 0.2 bar) | 5 μm |
06.5111 | Final Filter | 1400 L/hr. (at D p = 0.2 bar) | 0.45 μm |
06.5112 | Filter Insert, Final | 2800 L/hr. (at D p = 0.2 bar) | 0.45 μm |
50155514 | Prefilter Insert | 1200 L/hr. (at D p = 0.2 bar) | 5 μm |
06.5555 | Filter Insert, Sterile | 500 L/hr. (at D p = 0.2 bar) | 0.2 μm |
50155523 | Filter Insert, Sterile | 1000 L/hr. (at D p = 0.2 bar) | 0.2 μm |
06.5201 | Prefilter Insert | 750 L/hr. (at D p = 0.2 bar) | 5 μm |
Protect installations and systems from undissolved substances in water with these pre-filters, final filters and sterile filters. Thermo Scientific™ Point of Use Filter Inserts include 10- and 20-inch pre-filters, final filters and sterile filters for Point of Use systems.
Pre-filters 1μm, 5μm
Final filters
Sterile filters