Dionex™ NGES Next Generation Electrolytic Suppressor
Thermo Scientific™

Dionex™ NGES Next Generation Electrolytic Suppressor

Designed for a broad range of applications, the Thermo Scientific™ Dionex™ NGES Next Generation Electrolytic Suppressor offers excellent performance, particularly with smaller 4 μm particle-size ion chromatography columns.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityFor Use With (Application)Diameter
06000435 μeq/minCation analysis2 mm
060001200 μeq/minAnion analysis4 mm
060002100 μeq/minCation analysis4 mm
06000350 μeq/minAnion analysis2 mm
Catalog number 060004
Price (USD)
1,570.00
Each
Capacity:
35 μeq/min
For Use With (Application):
Cation analysis
Diameter:
2 mm
Price (USD)
1,570.00
Each

Meet the growing demands of ion chromatography (IC) with the Dionex NGES electrolytic suppressor. Housed in new hardware, this suppressor is designed for IC applications requiring high capacity, low noise, high backpressure resiliency, and fast startup. The Dionex NGES electrolytic suppressor is available in 2 mm and 4 mm formats.

Meet growing IC application demands with the Dionex NGES electrolytic suppressor

  • Full electrolytic regeneration without reagents
  • Sustains accurate results with exceptional peak resolution
  • Improved backpressure resilience, making it compatible with a wide range of post-suppressor devices and detectors, such as mass spectrometers

The Dionex NGES electrolytic suppressor is the recommended replacement for the Thermo Scientific™ Dionex™ DRS™ 600 Dynamically Regenerated Suppressor and the Thermo Scientific™ Dionex™ ERS™ 500 Electrolytically Regenerated Suppressor.

Software compatibility information

The Dionex NGES Suppressor requires a minimum version of the Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) to be recognized as “NGES” in the software. Please refer to the Dionex NGES Suppressor Manual or Installation Guide for the required minimum version for recognition and operation of a Dionex NGES Suppressor with your system. If your instrument is not using the required version of Chromeleon CDS or higher, please contact your local Thermo Fisher Scientific representative for additional assistance.

Specifications
Capacity35 μeq/min
Diameter2 mm
Dimensions (D x W x H)4.5 x 10.7 x 4.7 cm
Flow Rate0.75 mL/min max
For Use With (Application)Cation analysis
For Use With (Equipment)Dionex ion chromatography systems
OperationRecycle mode, External water mode (EWM)
Pressure500 psi backpressure max
SolventsMethanesulfonic acid and sulfuric acid eluents
Temperature Range15 to 50 °C
Void Volume< 15 μL
Weight (English)1.04 lb
Weight (Metric)470 g
TypeSuppressor
Unit SizeEach