Thermo Scientific™
Dionex™ ICS-900 Cation Consumable Bundles
Related applications:
Industrial Chromatography
Save time by ordering Thermo Scientific™ Dionex™ ICS-900 Cation Consumables Bundles, designed to enable the fast isocratic separation of lithium, sodium, ammonium, potassium, magnesium and calcium using methanesulfonic or sulfuric acid eluents in diverse sample matrices using the Dionex ICS-900 system.
Back to top
Convenient Consumables Bundle Simplifies Analysis
Compatible with organic solvents (except alcohols), high temperatures and high acid concentrations, the moderate capacity IonPac analytical columns in this bundle are the recommended replacement for all IonPac column applications.
This convenient consumables pack has everything required for your analysis, including specific analytical and guard columns, appropriate Dionex™ AMMS™ 300 Anion MicroMembrane™ Suppressor, application-specific Displacement Chemical Regeneration (DCR) bottle, eluent and regenerant concentrates. Bundle versions are also available without the eluent and regenerant concentrates, which are recommended for customers outside the United States.
Earlier instruments that utilize DCR, including the Dionex ICS-90A, ICS-90, DX-80 systems are compatible with this bundle.