Thermo Scientific™
Dionex™ ICS-900 Anion Consumable Bundles
 Related applications:
Industrial Chromatography
For simplicity and ease of use, Thermo Scientific™ Dionex™ ICS-900 Anion Consumable Bundles are designed to provide everything you need for the fast isocratic separation of inorganic anions from diverse and complex matrices such as drinking water, wastewater, food and beverages and scrubber solutions. The included Dionex™ IonPac™ columns meet or exceeds requirements of U.S EPA Method 300.0 A. Note, the IonPac Carbonate Eluent Anion-Exchange Column, an advanced carbonate eluent column, is recommended for fast analysis of inorganic anions.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Convenient Consumables Bundle Simplifies Analysis
Available in either 3mm or 4mm sizes, the included high-capacity IonPac Carbonate Eluent Anion-Exchange Column enables compliance with the requirements of U.S EPA Method 300.0 A and separates common inorganic anions in 13 minutes or less with a simple carbonate/bicarbonate eluent.
For faster separation of inorganic anions using a carbonate/bicarbonate eluent, the IonPac AS22-Fast column is recommended.
Bundle versions are also available without the eluent and regenerant concentrates, which are recommended for customers outside the United States.
Documents
Manuals & protocols