Samco™ Bio-Tite™ Specimen Containers - 20mL/35mm
Samco™ Bio-Tite™ Specimen Containers - 20mL/35mm
Samco™ Bio-Tite™ Specimen Containers - 20mL/35mm
Samco™ Bio-Tite™ Specimen Containers - 20mL/35mm
Thermo Scientific™

Samco™ Bio-Tite™ Specimen Containers - 20mL/35mm

Save valuable lab space with these specimen containers, which are stackable for easy storage. Containers have high-clarity for easy sample identification.
Have Questions?
Catalog NumberCapacity (Metric)
060181
also known as 06 0181
20 mL
Catalog number 060181
also known as 06 0181
Price (USD)
457.66
Online Exclusive
518.67
Save 61.01 (12%)
Case of 1000
Capacity (Metric):
20 mL
Price (USD)
457.66
Online Exclusive
518.67
Save 61.01 (12%)
Case of 1000

Save valuable lab space with Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers, which are stackable for easy storage. These shatter-resistant containers are manufactured from virgin polypropylene and have a proprietary, all plastic, leak-resistant seal that prevents bio-hazardous spills. Bio-Tite containers are tested at varying pressures, making them ideal for use in various transport systems.

  • Bio-Tite caps create leak-resistant seal for superior safety and handling
  • Dual-threaded caps create improved sealing
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
Specifications
Capacity (Metric)20 mL
Closure ColorWhite
Closure Size35 mm
DescriptionCap and container bagged separately, no label, white caps, bulk; Case of 1000
Diameter (Metric)35 mm
LabelNo Label
No. per Case1000/Cs.
No. per Pack1000/Pk.
Closure IncludedYes
MaterialVirgin Polypropylene
Product TypeSpecimen Container
ShapeCylindrical
SterilityNon-sterile
Unit SizeCase of 1000