Thermo Scientific™
Dionex™ ED50A Electrochemical Detector Cells
Catalog number: 060357
Thermo Scientific™
Dionex™ ED50A Electrochemical Detector Cells
Catalog number: 060357
Detect a variety of electro-active species using Thermo Scientific™ Dionex™ ED50A Electrochemical Detector Cells.
Catalog Number
060357
Unit Size
Each
Type
Electrochemical Detector Cell
Price (USD)
Full specifications
For Use WithAS50 Autosampler/TC/CC
OperationUsed for the detection of a variety of electro-active species.
TypeElectrochemical Detector Cell
DescriptionFor AS50,TC/CC
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Electrochemical Detector Cell
Price (USD)
Full specifications
For Use WithLC25 Chromatography Oven
OperationUsed for the detection of a variety of electro-active species.
TypeElectrochemical Detector Cell
Unit SizeEach
Catalog Number
060358
also known as 60358
Unit Size
Each
Type
Electrochemical Detector Cell
Price (USD)
Full specifications
For Use WithLC10/20/30
OperationUsed for the detection of a variety of electro-active species.
TypeElectrochemical Detector Cell
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeTypePrice (USD)
060357Full specifications
EachElectrochemical Detector CellRequest A Quote
For Use WithAS50 Autosampler/TC/CC
OperationUsed for the detection of a variety of electro-active species.
TypeElectrochemical Detector Cell
DescriptionFor AS50,TC/CC
Unit SizeEach
060356Full specifications
EachElectrochemical Detector CellRequest A Quote
For Use WithLC25 Chromatography Oven
OperationUsed for the detection of a variety of electro-active species.
TypeElectrochemical Detector Cell
Unit SizeEach
060358
also known as 60358
Full specifications
EachElectrochemical Detector CellRequest A Quote
For Use WithLC10/20/30
OperationUsed for the detection of a variety of electro-active species.
TypeElectrochemical Detector Cell
Unit SizeEach
Showing 3 of 3
Detects carbohydrates, amino acids (on a gold working electrode), cyanide, bromide, iodide, thiocyanate, sulfide, thiosulfate (on a silver electrode), alcohols, aldehydes, ketones, carboxylic acids (on a platinum electrode) and catecholamines. Also, detects S-containing amino acids and electroactive DNA bases and derivatives (on a carbon electrode).