Thermo Scientific™

Dionex™ RFC-30 Reagent-Free™ Controllers

Catalog number: 060668
Thermo Scientific™

Dionex™ RFC-30 Reagent-Free™ Controllers

Catalog number: 060668
Increase baseline stability, sensitivity and reproducibility while simplifying workflow with electrolytic eluent generation and suppression for the legacy Thermo Scientific™ Dionex™ ion chromatography (IC) system. Dionex™ RFC-30 Reagent-Free™ Controllers upgrade an existing IC instrument to a Dionex™ Reagent-Free™ IC (RFIC™) system to eliminate manual eluent preparation and focus on what matters— applications and results.
 
Catalog Number
060668
Unit Size
Each
Description
RFC-30 Reagent-Free Controller with EGC-MSA Cartridge
Price (USD)
Full specifications
For Use With (Application)Ion Chromatography
Flow Rate0.01 to 3 mL/min.
For Use With (Equipment)Dionex Atlas Electrolytic Suppressor or Self-Regenerating Suppressor; Dionex DX-120, DX-320, DX-500, DX-600, or ICS-2500 IC System
Max Eluent Concentration100 mM
Max. Pressure21 MPa (3000 psi)
TypeReagent-Free Controller
Min. Eluent Concentration0.1 mM
DescriptionRFC-30 Reagent-Free Controller with EGC-MSA Cartridge
Unit SizeEach
Catalog Number
Unit Size
Each
Description
RFC-30 Reagent-Free Controller with EGC-KOH Cartridge
Price (USD)
Full specifications
For Use With (Application)Ion Chromatography
Flow Rate0.01 to 3 mL/min.
For Use With (Equipment)Dionex Atlas Electrolytic Suppressor or Self-Regenerating Suppressor; Dionex DX-120, DX-320, DX-500, DX-600, or ICS-2500 IC System
Max Eluent Concentration100 mM
Max. Pressure21 MPa (3000 psi)
TypeReagent-Free Controller
Min. Eluent Concentration0.1 mM
DescriptionRFC-30 Reagent-Free Controller with EGC-KOH Cartridge
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
060668Full specifications
EachRFC-30 Reagent-Free Controller with EGC-MSA CartridgeRequest A Quote
For Use With (Application)Ion Chromatography
Flow Rate0.01 to 3 mL/min.
For Use With (Equipment)Dionex Atlas Electrolytic Suppressor or Self-Regenerating Suppressor; Dionex DX-120, DX-320, DX-500, DX-600, or ICS-2500 IC System
Max Eluent Concentration100 mM
Max. Pressure21 MPa (3000 psi)
TypeReagent-Free Controller
Min. Eluent Concentration0.1 mM
DescriptionRFC-30 Reagent-Free Controller with EGC-MSA Cartridge
Unit SizeEach
060667Full specifications
EachRFC-30 Reagent-Free Controller with EGC-KOH CartridgeRequest A Quote
For Use With (Application)Ion Chromatography
Flow Rate0.01 to 3 mL/min.
For Use With (Equipment)Dionex Atlas Electrolytic Suppressor or Self-Regenerating Suppressor; Dionex DX-120, DX-320, DX-500, DX-600, or ICS-2500 IC System
Max Eluent Concentration100 mM
Max. Pressure21 MPa (3000 psi)
TypeReagent-Free Controller
Min. Eluent Concentration0.1 mM
DescriptionRFC-30 Reagent-Free Controller with EGC-KOH Cartridge
Unit SizeEach
Showing 2 of 2
The Advantages of Reagent-Free IC
  • Time and labor savings with freedom from manual eluent preparation
  • Elimination of variability and potential contamination from manually produced eluent
  • Improved consistency and reproducibility thanks to precise control of eluent concentration
  • Greater sensitivity with lower background noise
  • Simplified workflow with convenient eluent cartridges
  • The Dionex RFC-30 Reagent-Free Controller Comes With:
  • The Reagent-Free Controller stand-alone unit with front panel control
  • EGC-KOH Cartridge or EGC-MSA Cartridge
  • CR-ATC Continuously Regenerated Anion Trap or CR-CTC Continuously Regenerated Cation Trap column
  • Cartridge holder with degas assembly
Note: Your choice of analytical column, guard column, and suppressor is required for use.

The Dionex RFC-30 Reagent-Free Controller provides current (1mA resolution) for a Dionex™ Atlas™ Electrolytic Suppressor (AES) or Self-Regenerating Suppressor (SRS™), current for an Eluent Generator Cartridge and current for a CR-TC Continuously Regenerating Trap Column.

Compatible Legacy IC Systems
  • Dionex™ DX-120, DX-320, DX-500, DX-600, or ICS-2500 IC system
  • Note: Use of the Dionex RFC-30 Reagent-Free Controller with the Dionex DX-120 system also requires the DX-120 Adapter Cable (part number 057861)
Configurations
  • Dionex RFC-30 Reagent-Free Controller with EGC-KOH Cartridge and CR-ATC Continuously Regenerated Anion Trap (part number 060667)
  • Dionex RFC-30 Reagent-Free Controller with EGC-MSA Cartridge and CR-CTC Continuously Regenerated Cation Trap (part number 060668)

Figures

Frequently asked questions (FAQs)

Citations & References