Water Softener Accessories
Water Softener Accessories
Actual product may vary
Thermo Scientific™
Water Softener Accessories
Remove hard ions, such as calcium and magnesium, using these accessories. Softened water extends the life of the reverse osmosis membrane.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
061000Hardness Test Kit
061160Mounting Block w/ Blending Valve 1 in.
062000Salt Tablets, 25 kg
062001Bacteria Guard
063998Sensor Controller 2 in.
063999Sensor Controller 1 in.
063997Sensor Controller 1.5 in.
150111Blending Valve, 1 in.
150110Blending Valve, 1.5 in.
Catalog number 061000
Price (CLP)
-
Type:
Hardness Test Kit
Remove hard ions, such as calcium and magnesium, using Thermo Scientific™ Water Softener Accessories. Softened water extends the life of the reverse osmosis membrane.
Sensor controller for water softening monitoring
  • The Sensor controller SE is a monitoring system for a quality-controlled operation of the water softening system
Total hardness measurement (titration solution)
  • Necessary to determine the feed water hardness and calculate the softener program settings
Bacteria guard for water softeners
  • For disinfecting the ion exchange bed during each regeneration
  • The bacteria guard extracts chlorine from a part of the salt water to disinfect the ion exchange resin
Flexible connecting hose 1in. with stainless steel jacketing, length 1 meter
  • Smart and easy connection of your Thermo Scientific Water softener to wall connectors
  • Alternative instead of fixed piping, which offers more flexibility for maintenance
Mounting block 1in. with blending valve
  • For comfortable bypassing of the softener for maintenance, reparations, etc
Blending valve OVP for 1in. softening systems for permanent hardness adjustment
  • If a specific water hardness is required after the softener, the blending valve will some hard water into the soft water, coming out of the softeners output
Blending valve OVP for 1 1/2in. softening systems for permanent hardness adjustment
  • If a specific water hardness is required after the softener, the blending valve will some hard water into the soft water, coming out of the softeners output
Specifications
For Use With (Equipment)Cabinet Softening System WAE, Double Softening System WAT, Double Softening System WMS
TypeHardness Test Kit
Unit SizeEach