Potassium hexachlororhenate(IV), 99.9% (metals basis)
Potassium hexachlororhenate(IV), 99.9% (metals basis)
Potassium hexachlororhenate(IV), 99.9% (metals basis)
Thermo Scientific Chemicals

Potassium hexachlororhenate(IV), 99.9% (metals basis)

CAS: 16940-97-9 | Cl6K2Re | 477.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 062113.06
also known as 062113-06
Price (USD)
650.65
Online Exclusive
723.00
Save 72.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
650.65
Online Exclusive
723.00
Save 72.35 (10%)
Each
Chemical Identifiers
CAS16940-97-9
IUPAC Namedipotassium hexachlororheniumdiuide
Molecular FormulaCl6K2Re
InChI KeyHGCRXIXEXSEJGV-UHFFFAOYSA-H
SMILES[K+].[K+].Cl[Re--](Cl)(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Potassium hexachlororhenate is used in preparation of supported rhenium tricarbonyls which catalyze propylene hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only