Dionex™ IonPac™ CS18 IC Columns
Actual product may vary
Dionex™ IonPac™ CS18 IC Columns
Dionex™ IonPac™ CS18 IC Columns
Thermo Scientific™

Dionex™ IonPac™ CS18 IC Columns

Green features
Low capacity cation-exchange column for gradient separation of polyvalent and moderately hydrophobic amines in complex matrices using aqueous eluents and elevated temperature.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Column Type
062878250 mmAnalytical Column
06288050 mmGuard Column
Catalog number 062878
Price (USD)
1,930.00
Each 1
Length (Metric):
250 mm
Column Type:
Analytical Column
Price (USD)
1,930.00
Each 1

Separate polar amines with the Thermo Scientific™ Dionex™ IonPac™ CS18 Cation-Exchange Column. This low-capacity, carboxylate-functionalized column is specifically optimized for the separation of common cations, small polar amines, and moderately hydrophobic and polyvalent anions using simple aqueous eluents and elevated temperatures. However, the newer Thermo Scientific™ Dionex™ IonPac™ CS19 or Dionex IonPac CS19-4μm Cation Exchange Columns are the recommended columns for these applications.

Cation and Amine Separations Using Suppressed and Non-suppressed Conductivity Detection
  • Low-capacity column optimized for the isocratic and gradient separations of polar amines, moderately hydrophobic amines, and polyvalent cations
  • Can be used with suppressed and nonsuppressed conductivity detection
  • Nonsuppressed conductivity detection is recommended to extend calibration linearity for ammonium or weak bases
  • Can also be used for many of the nonsuppressed applications supported by the Dionex IonPac SCS-1 column but with much greater column stability
  • Optimized for simple acidic gradient separations with minimal baseline shift
  • Low capacity facilitates modest acid concentrations to elute polyvalent cations
  • Compatible with moderate amounts of organic solvents, excluding alcohols, to enhance analyte solubility, modify column selectivity, and facilitate effective column clean-up
Formats Available for Application Flexibility
  • 2x250mm analytical columns
  • 2x50mm guard columns
  • Longer 250mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity290 μeq
Column ChemistryAcidic
Crosslinking55% DVB
DescriptionCS18; 2 x 250 mm
Flow Rate0.25 to 0.35 mL/min.
For Use With (Application)Cation Exchange
For Use With (Equipment)RFIC Systems
Green FeaturesFSC
HydrophobicityMedium
Max. Pressure4000 psi (275 bar)
Packing MaterialEthylvinylbenzene/Divinylbenzene
Particle Size6 μm
QuantityEach 1
Solvent CompatibilityAcidic Eluents (pH 0 to 7), 0 to 20% HPLC Solvents, Alcohols Should be Avoided
Solvents0 to 20% HPLC Solvents, Alcohols Should be Avoided.
Stationary PhaseIonPac CS18
Column TypeAnalytical Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)250 mm
pH0 to 7
Unit SizeEach 1