Dionex™ IonPac™ AS19 IC Columns
Actual product may vary
Dionex™ IonPac™ AS19 IC Columns
Dionex™ IonPac™ AS19 IC Columns
Dionex™ IonPac™ AS19 IC Columns
Dionex™ IonPac™ AS19 IC Columns
Dionex™ IonPac™ AS19 IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS19 IC Columns

Green features
Measure trace bromate and other oxyhalides in drinking water, ground water, wastewater, and other diverse sample matrices.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
062886250 mm2 mmAnalytical Column
062885250 mm4 mmAnalytical Column
06288750 mm4 mmGuard Column
06288850 mm2 mmGuard Column
072064250 mm0.4 mmAnalytical Column
07206550 mm0.4 mmGuard Column
Catalog number 062886
Price (USD)
1,930.00
Each
Length (Metric):
250 mm
Diameter (Metric):
2 mm
Column Type:
Analytical Column
Price (USD)
1,930.00
Each
Measure trace bromate and other oxyhalides in drinking water, ground water, wastewater, and other diverse sample matrices with Thermo Scientific™ Dionex™ IonPac™ AS19 Capillary, Analytical & Guard Columns. The column is specifically designed and recommended for determination of trace bromate, a toxic byproduct of ozone disinfection. The column's high capacity and selectivity facilitate the determination of bromate in drinking water at the low-μg/L level. The Dionex IonPac AS19 columns meet the performance requirements of U.S. EPA Methods 300.0 and 300.1.
Reduce Operating Costs With our New Capillary Column

The Dionex IonPac AS19 Capillary column is packed with the same material as the equivalent standard bore version (providing the same performance as a 4 mm column), but requires only 1/100th the eluent flow rate. The capillary format provides the advantage of significantly reduced eluent consumption, which helps reduce operating costs.

Superior Chromatographic Performance
  • High capacity: 240μeq per column (4 x 250 mm)
  • Operates at ambient or elevated temperatures with column selectivity optimized for 30°C
  • Compatible with organic solvents
  • Low background signal and enhanced analyte sensitivity when using the Thermo Scientific™
  • Dionex™ Anion Self-Regenerating Suppressor (ASRS™ 300) and a Reagent-Free™ (RFIC™) system
  • Analyze a Wide Range of Oxyhalides and Inorganic Anions
  • Oxyhalides and common inorganic anions including fluoride, chlorite, bromate, chloride, nitrite, bromide, chlorate, nitrate, phosphate, and sulfate in drinking water, groundwater, wastewater, and other diverse sample matrices.
  • Analyzes most drinking water without the use of sample pretreatment or preconcentration.
  • Ideal when used with a Reagent-Free™ Ion Chromatography (RFIC)™ system for automatic eluent generation
Designed for Detection of Trace Bromate

The key application for this column is the determination of trace bromate in drinking water matrices using a potassium hydroxide gradient with suppressed conductivity detection. The column's selectivity ensures that bromate can be quantified at low μg/L concentrations using suppressed conductivity detection even in the presence of very high concentrations of chloride, sulfate, and carbonate.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Anion Exchange Capacity60 μeq
Certifications/ComplianceMeets The Performance Requirements of US EPA Methods 300.0 and 300.1
Column Capacity60 μeq
Column ChemistryHydroxide
Column ConstructionPEEK
Crosslinking55% DVB
Flow Rate0.25 to 0.5 mL/min.
For Use With (Application)Analysis of Trace Bromate in Drinking Water
For Use With (Equipment)ASRS 300 with The IonPac AS19 Column for Eluent Suppression
Green FeaturesFSC
HydrophobicityLow
Ion Exchange GroupAlkanol Quaternary Ammonium Ion
Max. Pressure3000 psi (207 bar)
Packing MaterialPolymeric
Particle Size7.5 μm
Pore Size2000 &‌Aring;
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Stationary PhaseIonPac AS19
Column TypeAnalytical Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)250 mm
Unit SizeEach