Dionex™ IonPac™ AS22 IC Columns
Actual product may vary
Dionex™ IonPac™ AS22 IC Columns
Dionex™ IonPac™ AS22 IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS22 IC Columns

Green features
High capacity, anion-exchange column using an isocratic carbonate/bicarbonate eluent to separate common inorganic anions for compliance monitoring in diverse matrices.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
06413550 mm2 mmGuard Column
064137250 mm2 mmAnalytical Column
064141250 mm4 mmAnalytical Column
06413950 mm4 mmGuard Column
Catalog number 064135
Price (USD)
631.00
Each
Length (Metric):
50 mm
Diameter (Metric):
2 mm
Column Type:
Guard Column
Price (USD)
631.00
Each

Designed specifically for compliance monitoring of inorganic anions in accordance with U.S. EPA Methods 300.0 (A), the Thermo Scientific™ Dionex™ IonPac™ AS22 IC Columns provide fast analysis of drinking water, ground water and wastewater. With the Dionex IonPac AS22 column, using a simple isocratic carbonate/bicarbonate eluent, common inorganic anions can be quickly separated in a variety of sample matrices including drinking water, wastewater, process streams, and scrubber solutions.

Analysis of Common Inorganic Anions and Low Molecular Weight Organic Acids in Diverse Matrices
  • Compliance monitoring of common anions in drinking water as specified in U.S. EPA Method 300.0 (A)
  • Fast isocratic separation of common inorganic anions in simple matrices in 8 min.
  • Isocratic separation of common inorganic anions in complex matrices in 12 min.
  • Excellent retention of fluoride from the water dip and resolution of fluoride, acetate, and formate
  • Excellent resolution of the carbonate peak away from peaks for common inorganic anions
  • Ideal alternative, using carbonate/bicarbonate eluents, for IonPac AS4A-SC, AS12A, AS14 and AS14A inorganic anion applications.
  • Solvent compatibility permits easy column cleanup after the analysis of samples with hydrophobic components.

Multiple Formats Available for Application Flexibility

  • 4 x 250 mm, 2 x 250 mm analytical columns
  • 4 x 50mm, 2 x 50mm guard columns
  • Longer 250 mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs

Note: For simple, accurate eluent preparation, use the IonPac AS22 Eluent Concentrate.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceU.S. EPA Method 300.0 (A)
Column ChemistryCarbonate/Bicarbonate
For Use With (Application)Anion Exchange
For Use With (Equipment)IonPac AS22 Column
Green FeaturesFSC
Max. Pressure3000 psi (207 bar)
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Column TypeGuard Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)50 mm
Unit SizeEach