Thermo Scientific™
Dionex™ ICS-6000 Carbohydrate Removal Accessory Kit for ED Electrochemical Detection
 Related applications:
Industrial Chromatography
Eliminate labor-intensive sample cleanup and ensure reliable amino acid quantification by removing carbohydrates from amino acid samples in-line using the Thermo Scientific™ Dionex™ ICS-6000 Carbohydrate Removal Accessory Kits for ED Electrochemical Detection. The syringe pump and six-port valve in this kit work in conjunction with Thermo Scientific™ Dionex™ CRC Carbohydrate Removal Cartridges to enable automatic pretreatment of your amino acid samples on a Thermo Scientific™ Dionex™ AAA-Direct™ ion chromatography system.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
The kit's syringe pump and six-port valve is designed for the Thermo Scientific™ Dionex™ ICS-5000+/ICS-6000 DP Dual Pump, or the Dionex ICS-5000+/ICS-6000 SP Single Pump system and a Dionex Carbohydrate Removal Cartridge. Installation requires modification of the Dionex AAA-Direct system to connect the syringe pump to the autosampler, and redirect the pump and Dionex™ AminoPac PA™ 10 to the six-port switching valve.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Documents
Manuals & protocols