Dionex™ ED Electrochemical Detector Disposable Electrodes
Actual product may vary
Thermo Scientific™
Dionex™ ED Electrochemical Detector Disposable Electrodes
Employ the latest innovation in electrochemical detection, providing a new level of reproducibility and ease of use with Thermo Scientific™ Dionex™ Electrochemical Detector Disposable Electrodes for ED, ED50A, ED50 and ED40 amperometry cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialIncludesQuantity
066480Gold on PTFE2 gaskets (0.002 in.)6/Pk.
060082Gold on Polyester2 gaskets (0.002 in.)6/Pk.
060140Gold on Polyester4 x 2 gaskets (0.002 in.)4 x 6/Pk.
060139Gold on Polyester2 gaskets (0.002 in.)6/Pk.
060216Gold on Polyester4 x 2 gaskets (0.002 in.)4 x 6/Pk.
063003Silver6 gaskets (0.002 in.)6/Pk.
064440Platinum2 gaskets (0.002 in.)6/Pk.
069336Carbon on PEEK6 gaskets (0.002 in.)6/Pk.
Catalog number 066480
Price (USD)
756.00
Each 1
Material:
Gold on PTFE
Includes:
2 gaskets (0.002 in.)
Quantity:
6/Pk.
Price (USD)
756.00
Each 1
Employ the latest innovation in electrochemical detection, providing a new level of reproducibility and ease of use with Thermo Scientific™ Dionex™ Electrochemical Detector Disposable Electrodes for Electrochemical amperometry cells. They are less expensive than conventional, non-disposable electrodes and can be replaced more often without electrode reconditioning by polishing and other methods.
  • Disposable electrodes provide reproducible analyses, electrode to electrode and lot to lot, while maintaining good peak shape, high efficiency and excellent sensitivity
  • At the end of the recommended lifetime, or if the electrode becomes fouled, a new electrode is easily installed - polishing is a thing of the past
Four types of disposable electrodes to detect a wide variety of electroactive analytes
  • Gold working electrodes for carbohydrates and amino acids
  • Silver working electrode for cyanide
  • Platinum working electrode for alcohols
  • Carbon working electrode for S-containing amino acids
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionGold on PTFE Disposable Electrode
For Use With (Application)Electrochemical Detection
For Use With (Equipment)Electrochemical Detector
Includes2 gaskets (0.002 in.)
MaterialGold on PTFE
Product LineDionex
Quantity6/Pk.
TypeDisposable Electrode
Unit SizeEach 1