Product Image
Thermo Scientific™

Dionex™ Haloacetic Acid Internal Standards

Simplify the challenge of determining haloacetic acids (HAA) in drinking water with these convenient, pre-prepared standards IC analysis.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDescripción
069409Patrón interno de ácido tricloroacético Dionex, 1000 mg/l
069406Patrón interno de ácido monocloroacético Dionex, 1000 mg/l
069407Patrón interno de ácido monobromoacético Dionex, 1000 mg/l
069408Patrón interno de ácido dicloroacético Dionex, 1000 mg/l
Número de catálogo 069409
Precio (MXN)
-
Descripción:
Patrón interno de ácido tricloroacético Dionex, 1000 mg/l

Simplify the challenge of determining haloacetic acids (HAA) in drinking water with these convenient, pre-prepared standards IC analysis to meet U.S. EPA required minimum contamination levels. Designed to save time and eliminate errors in standards preparation, you can rely on Thermo Scientific™ Dionex™ Haloacetic Acid Internal Standards for purity and quality.

Three 1mL HAA internal standards with verified concentrations:

  • Monochloroacetic acid MCAA-2-13C, 1000mg/L
  • Monobromoacetic acid MBAA-1-13C, 1000mg/L
  • Trichloroacetic Acid TCAA-2-13C, 1000mg/L

Quality to Trust
All Thermo Scientific Dionex IC standards are traceable to National Institute of Standards and Technology (NIST™) Standard Reference Materials and are shipped with a Certificate of Analysis verifying the concentration.

Especificaciones
Concentración1000 mg/L
DescripciónPatrón interno de ácido tricloroacético Dionex, 1000 mg/l
Para utilizar con (aplicación)Detección de ESI/MS
RiesgosInflamable, irritante
FuncionamientoSe utiliza como un patrón interno.
CantidadEach
Componentes estándarÁcido tricloroacético, éter metil terbutílico
Concentración estándar1000 mg/l
Almacenamiento<10 °C
Volumen (métrico)1 mL
TipoPatrón interno
Unit SizeEach