Product Image
Thermo Scientific™
Dionex™ Automated Sample Preparation Valve Kits
Choose Thermo Scientific™ Dionex™ Automated Sample Preparation Valve Kits for use with ICS-1100/1600/2100 systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
069472Valve Kit (6-Port, 2 Position)
069473Valve Kit (Auxiliary)
Catalog number 069472
Price (USD)
-
Type:
Valve Kit (6-Port, 2 Position)
Choose Thermo Scientific™ Dionex™ Automated Sample Preparation Valve Kits for use with ICS-1100/1600/2100 systems.
  • Field installation
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)ICS-1100/1600/2100 Systems
OperationRequires field installation.
TypeValve Kit (6-Port, 2 Position)
Unit SizeEach