Acclaim™ Guard Holder and Coupler
Thermo Scientific™
Acclaim™ Guard Holder and Coupler
Green features
Use the Thermo Scientific™ Guard Holder and Coupler for your Thermo Scientific™ Acclaim™ Guard columns. They can be purchased separately or as a kit.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
069707Guard Cartridge Holder-Coupler Kit V-2
069580Guard Cartridge Holder V-2
074188Guard Cartridge Coupler V-2
Catalog number 069707
Price (USD)
433.38
Each
Product Type:
Guard Cartridge Holder-Coupler Kit V-2
Price (USD)
433.38
Each
Use the Thermo Scientific™ Guard Holder and Coupler for your Thermo Scientific™ Acclaim™ Guard columns. They can be purchased separately or as a kit.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Green FeaturesFSC
IncludesGuard column holder and coupler
QuantityEach
For Use With (Equipment)All Acclaim Guard Cartridges
Product LineAcclaim
Product TypeGuard Cartridge Holder-Coupler Kit V-2
Unit SizeEach