Tri-n-propyltin chloride
Tri-n-propyltin chloride
Tri-n-propyltin chloride
Thermo Scientific Chemicals

Tri-n-propyltin chloride

CAS: 2279-76-7 | C9H21ClSn | 283.427 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 071122.03
亦稱為 071122-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS2279-76-7
Molecular FormulaC9H23ClSn
InChI KeyZXVSRBCDRCJMBB-UHFFFAOYSA-N
SMILESCl.CCC[SnH](CCC)CCC
Molecular Weight (g/mol)285.44
規格Specification Sheet規格表
Appearance (Color)Colorless
Formliquid
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only