Di-n-butyltin dilaurate, 95%
Di-n-butyltin dilaurate, 95%
Di-n-butyltin dilaurate, 95%
Thermo Scientific™

Di-n-butyltin dilaurate, 95%

CAS: 77-58-7 | C32H64O4Sn
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 071130.14
also known as 071130-14
Price (EUR)
28,40
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
28,40
Each
Chemical Identifiers
CAS77-58-7
IUPAC Namedibutyltin dilaurate
Molecular FormulaC32H66O4Sn
InChI KeyXWVGXTYHPCKSLV-UHFFFAOYSA-N
SMILESCCCC[SnH2]CCCC.CCCCCCCCCCCC(O)=O.CCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride content0.15 wt%, max
Water Content (Karl Fischer Titration)0.5 %, max
Appearance (Color)Clear pale yellow to yellow
Assay (unspecified)17.7 to 18.9 % Total Tin
FormLiquid
Di-n-butyltin dilaurate is used as a catalyst for the production of polyurethanes as well as for the transesterification reactions. It is involved in the vulcanization of silicones and a stabilizer in polyvinyl chloride (PVC). It acts as a rust inhibitor for polyurethanes, polyols, silicones and as a fuel additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only