Diphenyltin dichloride, 90+%
Diphenyltin dichloride, 90+%
Diphenyltin dichloride, 90+%
Thermo Scientific Chemicals

Diphenyltin dichloride, 90+%

CAS: 1135-99-5 | C12H12Cl2Sn | 345.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 071139.18
also known as 071139-18
Price (EUR)
207,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
207,00
Each
Chemical Identifiers
CAS1135-99-5
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay from Supplier's CofA≥94%
Melting Point41 -43?C
Used in the manufacture of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only