Dimethyltin oxide
Dimethyltin oxide
Dimethyltin oxide
Thermo Scientific Chemicals

Dimethyltin oxide

CAS: 2273-45-2 | C2H6OSn | 164.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 071164.18
also known as 071164-18
Price (EUR)
671,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
671,00
Each
Chemical Identifiers
CAS2273-45-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Melting Point> 200?C (dec.)
Assay from Supplier's CofA≥94%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only