Tin(IV) chloride, 99.99% (metals basis)
Tin(IV) chloride, 99.99% (metals basis)
Tin(IV) chloride, 99.99% (metals basis)
Thermo Scientific Chemicals

Tin(IV) chloride, 99.99% (metals basis)

CAS: 7646-78-8 | Cl4Sn | 260.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 071203.22
also known as 071203-22
Price (TWD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7646-78-8
IUPAC Nametin(IV) chloride
Molecular FormulaCl4H4Sn
InChI KeyHTGSJGUTBKFNJC-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Tin(IV) chloride is a precursor to prepare organotin compounds such as tetralkyltin and dialkyldichlorotin(IV), which finds application as catalysts and polymer stabilizers. As a Lewis acid catalyst, it is used in Fridel-Crafts reactions for alkylation and cyclization. It is involved in the selective nitration of aromatic compounds in presence of fuming nitric acid. Furthermore, it is used to prepare tin(IV) oxide coating by sol-gel process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only