Thermo Scientific™
Dionex™ ICS-6000 IC Cube™
Catalog number: 072000
Thermo Scientific™
Dionex™ ICS-6000 IC Cube™
Catalog number: 072000
Instrument setup is a snap with the Thermo Scientific™ Dionex™ ICS-6000 IC Cube™. Requiring just half the connections of analytical systems, the Dionex ICS-6000 ion chromatography (IC) capillary system simplifies even dual-channel application prep by using two Dionex ICS-6000 IC Capillary Cubes, each providing precut tubing, color-coded connections, quick-to-install cartridge consumables and temperature-controlled column compartment.
Catalog Number
072000
Unit Size
Each
Description
Module with 4-port valve (no consumables)
Price (USD)
Full specifications
For Use With (Equipment)ICS-5000+/ICS-6000 Ion Chromatography Systems
TypeIon Chromatography Capillary System
Temperature Range15°C to 80°C
DescriptionModule with 4-port valve (no consumables)
Injection ValvesUp to two (one per cube) high-pressure valves, 4-port, 2-position. An optional 6-port, 2-position valve is available to support large loop and concentrator applications.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Module with 6-port valve (no consumables)
Price (USD)
Full specifications
For Use With (Equipment)ICS-5000+/ICS-6000 Ion Chromatography Systems
TypeIon Chromatography Capillary System
Temperature Range15°C to 80°C
DescriptionModule with 6-port valve (no consumables)
Injection ValvesUp to two (one per cube) high-pressure valves, 4-port, 2-position. An optional 6-port, 2-position valve is available to support large loop and concentrator applications.
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
072000Full specifications
EachModule with 4-port valve (no consumables)Request A Quote
For Use With (Equipment)ICS-5000+/ICS-6000 Ion Chromatography Systems
TypeIon Chromatography Capillary System
Temperature Range15°C to 80°C
DescriptionModule with 4-port valve (no consumables)
Injection ValvesUp to two (one per cube) high-pressure valves, 4-port, 2-position. An optional 6-port, 2-position valve is available to support large loop and concentrator applications.
Unit SizeEach
078841Full specifications
EachModule with 6-port valve (no consumables)Request A Quote
For Use With (Equipment)ICS-5000+/ICS-6000 Ion Chromatography Systems
TypeIon Chromatography Capillary System
Temperature Range15°C to 80°C
DescriptionModule with 6-port valve (no consumables)
Injection ValvesUp to two (one per cube) high-pressure valves, 4-port, 2-position. An optional 6-port, 2-position valve is available to support large loop and concentrator applications.
Unit SizeEach
Showing 2 of 2
Robust Performance Simple
  • Improved peak dispersion and signal-to-noise ratio due to minimum dead volumes
  • Quick-to-connect, color-coded labels and connections
  • Easy-to-insert and remove, cartridge-based consumables
  • Freedom from metal contamination with inert PEEK flowpath
Advanced Applications
Even dual-channel capillary IC is now simple to set up, as the Dionex ICS-6000 system holds two Dionex ICS-6000 IC Capillary Cubes with independently temperature-controlled column compartments. This enables a great variety of time-saving, dual-channel applications, e.g. anion analysis at 30°C and cation analysis in the second channel at 60°C.
Flexible Components
  • Set up a wide variety of low-baseline, high-sensitivity capillary IC applications quickly and easily with Dionex ICS-6000 IC Capillary Cube components:
  • High sensitivity and low detection limits using the Thermo Scientific™ Dionex™ ACES™ 300/CCES™ 300 Anion or Cation Capillary Electrolytic Suppressor
  • Flexible setup with the four-port, two -osition 0.4µL internal loop valve
  • Reduced background using the Dionex CRD Carbonate Removal Device
  • Stable baselines and lower detection limits thanks to the Eluent Degasser (EG Degas)
  • Protection for sensitive samples with temperature-controlled guard and separation column compartments