Dionex™ ICS-6000/4000 ED Electrochemical Detector Cell
Thermo Scientific™
Dionex™ ICS-6000/4000 ED Electrochemical Detector Cell
The ICS-6000/4000 ED Electrochemical Detector Cell is for use with the ICS-6000 and ICS-4000 ion chromatography systems.
Have Questions?
Catalog NumberFor Use With (Equipment)
072044ICS-5000+/ICS-6000, ICS-4000 Electrochemical Detectors
Catalog number 072044
Price (USD)
-
For Use With (Equipment):
ICS-5000+/ICS-6000, ICS-4000 Electrochemical Detectors
Use the Thermo Scientific™ Dionex™ ICS-6000/4000 ED Electrochemical Detector Cell for ICS-6000 and ICS-4000 ion chromatography systems.
  • Cell body includes PEEK (Victrex plc) Yoke Block
Specifications
DescriptionElectrochemical Detector Cell
For Use With (Equipment)ICS-5000+/ICS-6000, ICS-4000 Electrochemical Detectors
Detector TypeElectrochemical (ECD)
TypeElectrochemical Detector Cell
Unit SizeEach