Dionex™ IonPac™ AS18-Fast IC Columns
Actual product may vary
Dionex™ IonPac™ AS18-Fast IC Columns
Dionex™ IonPac™ AS18-Fast IC Columns
Dionex™ IonPac™ AS18-Fast IC Columns
Dionex™ IonPac™ AS18-Fast IC Columns
Dionex™ IonPac™ AS18-Fast IC Columns
Dionex™ IonPac™ AS18-Fast IC Columns
Dionex™ IonPac™ AS18-Fast IC Columns
Dionex™ IonPac™ AS18-Fast IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS18-Fast IC Columns

Green features
Achieve fast analysis of inorganic anions and low-molecular-weight organic acids in environmental waters in less than 6 min using these high capacity, hydroxide-selective anion-exchange columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
072062150 mm0.4 mm
075759150 mm2 mmAnalytical Column
075760150 mm4 mmAnalytical Column
07576130 mm2 mmGuard Column
07576230 mm4 mmGuard Column
07206335 mm0.4 mmGuard Column
Catalog number 072062
Price (MXN)
-
Length (Metric):
150 mm
Diameter (Metric):
0.4 mm
Use these shorter 150 mm Thermo Scientific™ Dionex™ IonPac™ AS18-Fast columns for fast runs when monitoring inorganic anions in drinking water and wastewater samples for compliance with U.S. EPA Methods 300.0 (A) and 300.1. Dionex IonPac AS18 columns are high capacity hydroxide-selective anion-exchange columns. Having the same selectivity as the Dionex IonPac AS18 colum , the shorter format significantly reduces run times of common inorganic anions and low molecular weight organic acids in diverse sample matrices including drinking water, wastewater, leachates, foods, beverages, and pharmaceutical preparations.

Analysis of Common Inorganic Anions and Low-Molecular Weight Organic Acids in Diverse Matrices

  • Compliance monitoring of common anions in drinking water as specified in U.S. EPA Method 300.0 (A)
  • Determination of low levels of inorganic anions in complex matrices using large-loop injections
  • Superior retention and quantification of fluoride, acetate, and formate
  • Fast separation of the common inorganic anions in 5 min in simple matrices using 150 mm column
  • High efficiency, fast analysis use the 150 mm column for increased sample throughput
  • Compatible with organic solvents to enhance analyte solubility, modify column selectivity, or allow effective column cleanup
Recommended for Inorganic Anions in Diverse Sample Matrices
  • Source water and drinking water, municipal and industrial wastewater
  • Industrial cooling water and waste water, acid rain
  • Foods and beverages, anionic counterions in pharmaceutical preparations

Multiple Formats for Application Flexibility

The shorter 150 mm Dionex IonPac AS18-Fast columns produce faster runs of relatively clean matrices compared to the longer 250 mm columns and are available in three formats with guard columns:

  • 4 x 150 mm standard bore AS18-Fast columns with 4 x 30 mm guard columns
  • 2 x 150 mm microbore AS18-Fast columns with 2 x 30 mm guard columns
  • 0.4 x 150 mm capillary AS18-Fast columns with 0.4 x 35 mm guard columns

Capillary Columns Reduce Operating Costs from Lower Eluent Requirements

Capillary columns provide significantly reduced operating and waste disposal costs through eluent conservation. Packed with the same material, the Dionex IonPac AS18-Fast capillary columns provide the same performance as the standard bore and microbore counterparts. Operating costs are reduced because the capillary system operates at only 1% of the eluent flow rate of the standard bore system. Capillary columns are used with the Thermo Scientific™ Dionex™ ICS-4000 and ICS-5000+ Capillary systems.

Need Higher Resolution or Faster Separations

  • If you need higher resolution separations of more complex sample matrices, refer to the longer 250 mm Dionex™ IonPac™ AS18 Hydroxide-Selective Anion-Exchange column
  • For fastest, highest resolution separations of relatively cleaner sample matrices, refer to the small particle Dionex™ IonPac™ AS18-4μm Hydroxide-Selective Anion-Exchange column. Use of 4μm columns requires a high-pressure IC system.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Anion Exchange Capacity1.71 μeq
Certifications/ComplianceU.S. EPA Methods 300.0 (A) and 300.1
Column Capacity1.71 μeq
Column ChemistryHydroxide
Column ConstructionPEEK
Crosslinking55% DVB
Diameter (Metric) Latex65 nm
Flow Rate0.01 to 0.03 mL/min.
For Use With (Application)Anion Exchange
For Use With (Equipment)ICS-5000 Capillary IC System
Green FeaturesFSC
HydrophobicityLow
Ion Exchange GroupAlkanol Quaternary Ammonium Ion
Max. Pressure5000 psi (345 bar)
Packing MaterialEthylvinylbenzene/Divinylbenzene
Particle Size7.5 μm
Pore Size2000 &‌Aring;
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Solvents0 to 100% of HPLC Solvents
Stationary PhaseIonPac AS18-Fast
TypeAS18
Diameter (Metric)0.4 mm
FormatCapillary
Length (Metric)150 mm
pH0 to 14
Unit SizeEach 1