Dionex™ SRD-10 Suppressor Regenerant Detector
Thermo Scientific™
Dionex™ SRD-10 Suppressor Regenerant Detector
Protect your suppressor and make every run count; automatically stop your ion chromatography system if regenerant flow stops or slows.
Have Questions?
Catalog NumberQuantity
074395Each 1
Catalog number 074395
Price (USD)
856.00
Each
Quantity:
Each 1
Price (USD)
856.00
Each
Protect your suppressor and make every run count when using external water or chemical regenerant mode. The Thermo Scientific™ Dionex™ SRD-10 Suppressor Regenerant Detector automatically disables the eluent pump if the suppressor regenerant flow stops or slows, preventing damage to the suppressor or a wasted ion chromatography (IC) run.
Simple Peace of Mind
  • Easy-to-use, stand-alone device
  • Set-and-forget operation; continuously monitors regenerant flow and automatically shuts off the eluent pump via a TTL command if regenerant flow is restricted or stops
Protection Where You Need It
  • The Dionex™ SRS™ Self-Regenerating Suppressor is designed for use with these applications:
  • Dionex™ SRS™ 300 Self-Regenerating Suppressor operated in external water mode
  • Dionex™ MMS™ 300 MicroMembrane™ Suppressor operated in chemical regenerant mode
Prevent Suppressor Damage
In external water mode, if the water supply is stopped or depleted, the IC system can continue to operate and current can be applied to the suppressor even after regenerant flow stops. Unless the current is turned off, this will quickly cause irreversible damage to the Dionex SRS Self-Regenerating Suppressor. In chemical regenerant mode, running out of regenerant will not damage the Dionex MMS MicroMembrane Suppressor, but it will adversely affect the chromatographic run. In both cases, the Dionex SRD-10 Suppressor Regenerant Detector will stop the run and shut off the SRS suppressor if needed, preventing suppressor damage and saving eluent and samples.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionSuppressor regenerant detector
For Use With (Equipment)SRS 300 Self-Regenerating Suppressor Operated in External Water Mode
OperationStand-alone device that monitors liquid flow to a suppressor and automatically disables the eluent pump if the suppressor regenerant flow stops or slows.
QuantityEach 1
TypeSuppressor Regenerant Detector
Unit SizeEach