Thermo Scientific™
Dionex™ InGuard™ HRP Organics Removal Kit
 Related applications:
Industrial Chromatography
Use the Thermo Scientific™ Dionex™ InGuard™ HRP Organics Removal Kit for all fittings and consumables you will need to configure the Dionex InGuard HRP hydrophilic reversed-phase cartridge for automated removal of organic interferences from ion chromatography (IC) samples. This kit is particularly useful for processing dairy product samples or those contaminated with oil and grease.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Conveniently includes:
  • One package of four Dionex InGuard HRP Cartridges (9 x 24 mm)
  • One Thermo Scientific™ Dionex™ IonPac™ UTAC-XLP2 Ultra Trace Anion Concentrator Column (6 x 16 mm)
  • One Dionex IonPac ATC-HC Anion Trap Columns (9 x 75 mm)
  • 5 feet of PEEK tubing
  • 6 fittings
  • 10 ferrules
Documents
Manuals & protocols