Thermo Scientific™
Dionex™ InGuard™ Ag plus Na Halide Removal Kit
Related applications:
Industrial Chromatography
Use the Thermo Scientific™ Dionex™ InGuard™ Ag plus Na Halide Removal Kit for all fittings and consumables you will need to configure the Dionex InGuard Ag and Dionex InGuard Na cartridges for automated removal of halide from ion chromatography (IC) samples. This kit is particularly useful for removal of chloride from industrial or environmental brine samples.
Back to top
Conveniently includes:
  • One package of four Dionex InGuard Ag Cartridges (9 x 24 mm)
  • One package of four Dionex InGuard Na Cartridges (9 x 24 mm)
  • One Thermo Scientific™ Dionex™ IonPac™ UTAC-XLP2 Ultra Trace Anion Concentrator Column (6 x 16 mm)
  • One Dionex IonPac ATC-HC Anion Trap Columns (9 x 75 mm)
  • One inline filter
  • 5 feet of PEEK tubing
  • 10 fittings
  • 15 ferrules