Dionex™ AS-AP Autosampler Vial Kits
Actual product may vary
Thermo Scientific™
Dionex™ AS-AP Autosampler Vial Kits
Use Thermo Scientific™ Dionex™ AS-AP Autosampler Vial Kits offered in polystyrene, glass or polypropylene, with caps and split septa.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionMaterialIncludes
074927Single injection sealing disks, 10 mLPTFESealing disks only
055427Vial Kit, 1.5 mLGlassCaps and Split Septa
055428Vial Kit, 0.3 mLPolypropyleneCaps and Split Septa
079812Vial Kit, 1.5 mLPolypropyleneCaps and Split Septa
074228PolystyreneCaps and Blue Split Septa
055058PolystyreneCaps and Split Septa
Catalog number 074927
Price (MXN)
-
Description:
Single injection sealing disks, 10 mL
Material:
PTFE
Includes:
Sealing disks only
Use Thermo Scientific™ Dionex™ AS-AP Autosampler Vial Kits offered in polystyrene, glass or polypropylene, with caps and split septa.
Septa included are blue.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionSingle injection sealing disks, 10 mL
For Use With (Equipment)AS-AP Autosampler
IncludesSealing disks only
MaterialPTFE
QuantityPack of 100
TypeSingle injection sealing disks, 10 mL
Unit SizePack of 100