Dionex™ EG Eluent Generator Kits
Dionex™ EG Eluent Generator Kits
Actual product may vary
Thermo Scientific™
Dionex™ EG Eluent Generator Kits
Run ion chromatography gradients as easily as isocratic by installing the Thermo Scientific™ Dionex™ EG Cartridge Kits in the Dionex ICS-6000 EG Eluent Generator.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
075522Analytical EG KitDionex ICS-6000 EG Eluent Generator
074459Capillary EG Cartridge KitDionex ICS-5000+ IC System (Capillary)
22181-60202Capillary EG KitDionex ICS-6000 EG Eluent Generator
22181-60951Dionex RFIC+ Dual EGC Degasser Dionex ICS-6000 HPIC System in Dual EGC Mode
22181-62038RFIC+ Dual EGC Degasser Ship KitDionex ICS-6000 HPIC System in Dual EGC Mode
22181-62048RFIC+ Dual EGC Vacuum Degas KitDionex ICS-6000 HPIC System in Dual EGC Mode
106-60001RFIC Eluent DegasserDionex Inuvion IC System
Catalog number 075522
Price (MXN)
-
Description:
Analytical EG Kit
For Use With (Equipment):
Dionex ICS-6000 EG Eluent Generator
Achieve reproducible and reliable chromatography separations using Thermo Scientific™ Dionex™ Reagent-Free IC (RFIC™) technologies. Dionex Eluent Generator accessories allow you to operate your RFIC system with minimal effort through plug-and-play setup.
Operate your ion chromatography system with minimal effort when using Dionex EG Eluent Generator accessories.
Specifications
Amperage90-265 VAC 47-63 Hz
DescriptionAnalytical EG Kit
Flow Rate0.1 to 3.000 mL/min.
For Use With (Equipment)Dionex ICS-6000 EG Eluent Generator
IncludesAnalytical degasser module and tubing
Pressure5000 psi
SolventsMax. Solvent concentration - Cations: None; Anions: 25%
TypeAnalytical Eluent Generator Kit (Eluent Concentration: 0.01 to 100 mM)
Unit SizeEach