Thermo Scientific™
Rocket™ Synergy 2 Evaporation Systems
Catalog number: 075906
Thermo Scientific™
Rocket™ Synergy 2 Evaporation Systems
Catalog number: 075906
Concentrate or dry large-volume samples rapidly and in parallel using the automated Thermo Scientific™ Rocket™ Synergy 2 Evaporator System. Up to 6 x large-volume 450 mL flasks or 18 x 60 mL Thermo Scientific™ Dionex™ ASE™ vials can be simultaneously processed without user intervention. Minimize cross-contamination and sample loss by directly concentrating your samples into an autosampler vial. This eco-friendly system ensures high solvent recoveries even with volatile organic solvents.
Catalog Number
075906
Unit Size
Each
Type
Evaporator Chiller Connection Kit
Price (USD)
Full specifications
DescriptionChiller Connection Kit
Holds450 mL Flasks
TypeEvaporator Chiller Connection Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Rocket Synergy 2 Evaporator System (120V)
Price (USD)
Full specifications
TypeRocket Synergy 2 Evaporator System (120V)
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Rocket Synergy 2 Evaporator System (240V)
Price (USD)
Full specifications
TypeRocket Synergy 2 Evaporator System (240V)
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Rocket Synergy 2 Evaporator System with Inert Gas Purge (120V)
Price (USD)
Full specifications
TypeRocket Synergy 2 Evaporator System with Inert Gas Purge (120V)
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Rocket Synergy 2 Evaporator System with Inert Gas Purge (240V)
Price (USD)
Full specifications
TypeRocket Synergy 2 Evaporator System with Inert Gas Purge (240V)
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Evaporator Chiller (120 V)
Price (USD)
Full specifications
Voltage120 V
TypeEvaporator Chiller (120 V)
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Evaporator Chiller (240 V)
Price (USD)
Full specifications
Voltage240 V
TypeEvaporator Chiller (240 V)
Unit SizeEach
Catalog Number
Unit Size
Each
Type
PM Kit for Rocket Synergy 2 Systems
Price (USD)
Full specifications
TypePM Kit for Rocket Synergy 2 Systems
Unit SizeEach
Catalog Number
Unit Size
Each
Type
PM Kit for Rocket Synergy 2 IGP Systems
Price (USD)
Full specifications
TypePM Kit for Rocket Synergy 2 IGP Systems
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Evaporator Maintenance Service Kit
Price (USD)
Full specifications
DescriptionService Kit for Rocket Synergy 2 Evaporator Rotor
For Use With (Equipment)Rocket Synergy Evaporator Rotor
TypeEvaporator Maintenance Service Kit
Unit SizeEach
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeTypePrice (USD)
075906Full specifications
EachEvaporator Chiller Connection KitRequest A Quote
DescriptionChiller Connection Kit
Holds450 mL Flasks
TypeEvaporator Chiller Connection Kit
Unit SizeEach
C0960-01-00085Full specifications
EachRocket Synergy 2 Evaporator System (120V)Request A Quote
TypeRocket Synergy 2 Evaporator System (120V)
Unit SizeEach
C0960-01-00086Full specifications
EachRocket Synergy 2 Evaporator System (240V)Request A Quote
TypeRocket Synergy 2 Evaporator System (240V)
Unit SizeEach
C0960-01-00087Full specifications
EachRocket Synergy 2 Evaporator System with Inert Gas Purge (120V)Request A Quote
TypeRocket Synergy 2 Evaporator System with Inert Gas Purge (120V)
Unit SizeEach
C0960-01-00088Full specifications
EachRocket Synergy 2 Evaporator System with Inert Gas Purge (240V)Request A Quote
TypeRocket Synergy 2 Evaporator System with Inert Gas Purge (240V)
Unit SizeEach
075905Full specifications
EachEvaporator Chiller (120 V)Request A Quote
Voltage120 V
TypeEvaporator Chiller (120 V)
Unit SizeEach
076364Full specifications
EachEvaporator Chiller (240 V)Request A Quote
Voltage240 V
TypeEvaporator Chiller (240 V)
Unit SizeEach
C0960-01-00093Full specifications
EachPM Kit for Rocket Synergy 2 Systems
TypePM Kit for Rocket Synergy 2 Systems
Unit SizeEach
C0960-01-00094Full specifications
EachPM Kit for Rocket Synergy 2 IGP Systems
TypePM Kit for Rocket Synergy 2 IGP Systems
Unit SizeEach
00960-01-00058Full specifications
EachEvaporator Maintenance Service KitRequest A Quote
DescriptionService Kit for Rocket Synergy 2 Evaporator Rotor
For Use With (Equipment)Rocket Synergy Evaporator Rotor
TypeEvaporator Maintenance Service Kit
Unit SizeEach
Showing 10 of 10
Rapid and efficient solvent evaporation
  • Low temperature, low pressure steam heating technology facilitates fast evaporation and precise temperature control.
  • Five times faster than conventional sample evaporators.
  • Dri-Pure™ technology prevents solvent bumping and cross-contamination.
  • Genevac AutoStop feature allows automated end-point detection.
Easy to operate
  • Easy to use controls and software.
  • Walk-away automation.
  • Flip-flop system eliminates manual sample transfer by automating it from ASE vials.
  • SampleGenie™ enables samples to be concentrated or dried directly in storage or autosampler vials.
Enhanced laboratory productivity
  • Unattended evaporation/concentration of up to 18 x 60 mL ASE vials or 6 x large-volume 450 mL flasks.
  • Two-stage cold trap allows high solvent recovery.
  • New trolley enables evaporator portability within the lab.
Recommended use
  • Use in conjunction with the the Thermo Scientific™ Dionex™ ASE™ 350 Accelerated Solvent Extractor to optimize sample preparation workflow and to maximize laboratory productivity.
  • Analyze prepared samples with Thermo Scientific LC or GC systems for a fully integrated workflow.