Dionex™ ICS-5000+ DC Detector/Chromatography Compartment
Thermo Scientific™  Related applications: Industrial Chromatography
Improve stability and sensitivity with flexible and precise temperature control for your conductivity detector and columns. The Thermo Scientific™ Dionex™ ICS-5000+ DC Detector/Chromatography Compartment houses and organizes chromatography components such as valves, Dionex™ IC Cube™ module(s), as well as conductivity and electrochemical detectors and cells in a compact space that minimizes connection lengths.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Introducing shared lists
Back to top
Description
Available in Two Versions
  • Standard version for applications requiring an upper zone temperature of 18-40 °C
  • Low temperature for capillary applications requiring an upper zone temperature of 10-40 °C.
Three Convenient Sections for Automation, Detection, and Separation
  • Configure the automation section with two IC Cube modules or with the Thermo Scentific™ Dionex™ Automation Manager (for standard bore and microbore columns only)
  • Use up to two Thermo Scentific Dionex detectors in series or parallel
  • Employ up to six separate temperature zones simultaneously: separation section, detection section, two detectors and IC Cube modules or postcolumn reaction coils
Documents
Manuals & protocols