Dionex™ ICS-6000 AM Automation Manager
Thermo Scientific™
Dionex™ ICS-6000 AM Automation Manager
Simplify and automate ion chromatography sample preparation, preconcentration, matrix elimination, and postcolumn reagent additions in Dionex ICS-6000 HPIC systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
0759521 6-port High Pressure Valve, 1 Low Pressure 3-Way Valve
079833No valves installed
0759502 10-port High-Pressure Valves, 2 Low-Pressure 3-Way Valves
0759511 10-port High Pressure Valve, 1 Low Pressure 3-Way Valve
Catalog number 075952
Price (USD)
-
Includes:
1 6-port High Pressure Valve, 1 Low Pressure 3-Way Valve
Save time and streamline even complex ion chromatography (IC) applications, including automated sample preparation, preconcentration, matrix elimination, and postcolumn reagent addition. Configure the Thermo Scientific™ Dionex™ ICS-6000 AM Automation Manager in the upper compartment of the Dionex DC Detector/Chromatography Compartment with a choice of two high-pressure rotary valves or two low-pressure solenoid valves. Setup is easy with components recognized, configured and controlled through Thermo Scientific™ Chromeleon™ Chromatography Data System software.
  • Complete automation of even complex applications
  • Full support for standard bore and microbore chemistries on the Dionex™ ICS-6000 HPIC™ Standard and Microbore and Hybrid systems
  • Ability to streamline postcolumn reagent addition using Thermo Scientific™ Dionex™ AutoPrep™ and RFIC™-ESP options
Not available for capillary IC.
Specifications
For Use With (Equipment)Dionex ICS-5000+/ICS-6000 Ion Chromatography Systems with Chromeleon Software
Includes1 6-port High Pressure Valve, 1 Low Pressure 3-Way Valve
OperationStreamlines and automates ion chromatography (IC) applications.
TypeAutomation Manager
Unit SizeEach